Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombomodulin Mouse anti-Human,Rat, Unconjugated, Clone: 1009, MilliporeSigma™

Catalog No. MABS21562MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS21562MI 25 μL
MABS21561MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS21562MI Supplier MilliporeSigma Supplier No. MABS215625UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Thrombomodulin Monoclonal antibody specifically detects Thrombomodulin in Human,Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Inhibits Activity/Function, Radioimmunoassay, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thrombomodulin
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone 1009
Conjugate Unconjugated
Gene Accession No. NP_000352
Gene Alias TM;Fetomodulin;CD141
Gene Symbols THBD, THRM
Host Species Mouse
Immunogen Recombinant fragment corresponding to 241 amino acids corresponding to six EGF repeats from human Thrombomodulin.
Molecular Weight of Antigen ∼60.33 kDa calculated.
Purification Method Protein G Purified
Quantity 25 μL
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone 1009 is a mouse monoclonal antibody that detects Thrombomodulin.
Target Species Human, Rat
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.