Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Thrombopoietin Monoclonal Antibody (9H10H8D11), Invitrogen™

Catalog No. PIMA543476
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543476 50 μL
PIMA542987 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543476 Supplier Invitrogen™ Supplier No. MA543476
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specificity: Human THPO

Thrombopoietin (TPO, THPO, MPL ligand, megakaryocyte colony-stimulating factor) is responsible for the generation of platelets through its action on hematopoietic cells committed to the megakaryocyte lineage. Binding of thrombopoietin to its receptor, MPL (CD110), initiates a signaling cascade via JAK/STAT, MAPK, and PI3K/AKT signaling pathways that leads to the proliferation and differentiation of megakaryocytes and, ultimately, to the production of platelets (thrombopoiesis). Thrombopoietin is constitutively produced by the liver and has a predicted molecular weight of 38 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thrombopoietin
Applications ELISA
Classification Monoclonal
Clone 9H10H8D11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene THPO
Gene Accession No. P40225
Gene Alias C-mpl ligand; Megakaryocyte colony-stimulating factor; megakaryocyte growth and development factor; megakaryocyte stimulating factor; Mgdf; MKCSF; ML; MPL ligand; Mpllg; MSA; Myeloproliferative leukemia virus oncogene ligand; prepro-thrombopoietin; TDH2A; THCYT1; THPO; Thrombopoietin; thyroid microsomal antigen; thyroid peroxidase; thyroperoxidase; Tpo; TPX
Gene Symbols THPO
Host Species Mouse
Immunogen Human full-length cell-derived rhTHPO.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7066
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.