Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombospondin-1 Antibody (A6.1), Novus Biologicals™
SDP

Catalog No. p-7106100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002059SS 0.05 mL
NB1002059 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1002059SS Supplier Novus Biologicals Supplier No. NB1002059SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 7 publications

Thrombospondin-1 Monoclonal antibody specifically detects Thrombospondin-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Block/Neutralize.

Specifications

Antigen Thrombospondin-1
Applications Blocking Assay, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone A6.1
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:25-1:50, Immunohistochemistry-Paraffin 1:25-1:50, Immunohistochemistry-Frozen, Blocking Assay
Formulation PBS (pH 7.4), 0.2% BSA, Tween-20 with 0.05% Sodium Azide
Gene Accession No. P07996
Gene Alias THBS, THBS-1, thrombospondin 1, thrombospondin-1p180, TSP1thrombospondin-1, TSPTSP-1
Gene Symbols THBS1
Host Species Mouse
Immunogen BALB/C mice were injected with reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets.
Purification Method Protein A or G purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7057
Test Specificity This is specific to 450 kD protein, a non-reduced form, and 170 to 180 kD protein, a reduced form, of Thrombospondin.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Sheep
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.