Learn More
Thrombospondin 1 Monoclonal Antibody (A6.1), Invitrogen™
Catalog No. PIMA513398
Catalog No. PIMA513398 Supplier Thermo Scientific Supplier No. MA513398
Add to Cart
Add to Cart
Mouse Monoclonal Antibody
MA5-13398 targets Thrombospondin in FACS, ICC/IF, IHC (P), IM, IP, and WB applications and shows reactivity with Bovine, Canine, Equine, Human, mouse, Ovine, Porcine, and Rat samples. The MA5-13398 immunogen is reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets.
Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.