Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Thrombospondin 1 Monoclonal Antibody (A65M)
GREENER_CHOICE

Catalog No. CSI0026502
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CSI0026502 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CSI0026502 Supplier Invitrogen™ Supplier No. CSI0026502
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CSI 002-65-02 is highly specific for the low-calcium conformation of thrombospondin. There is no evidence for cross-reactivity with other connective tissue proteins (vitronectin, fibronectin, elastin, collagen, laminin). CSI 002-65-02 cross-reacts with human thrombospondin. Other species have not been tested. CSI 002-65-02 has been successfully used in ELISA, Immunohistochemistry and Immunoprecipitation procedures. CSI 002-65 can also be used to probe for the low-calcium conformation of thrombospondin. The CSI 002-65-02 epitope is only present in thrombospondin prepared with low levels of calcium. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.

Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thrombospondin 1
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone A65M
Concentration 0.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene THBS1
Gene Accession No. P07996, Q28178
Gene Alias glycoprotein G; tbsp1; THBS; Thbs1; Thbs-1; thrombospondin; thrombospondin 1; thrombospondin-1; thrombospondin-1p180; thrombospondin-p50; ThrombospondinTHBS1; TSP; TSP1; TSP-1
Gene Symbols THBS1
Host Species Mouse
Immunogen Extracellular matrix materila fom cultured bovine corneal endothelial cells.
Purification Method Protein A/G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 281530, 7057
Target Species Human, Bovine
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.