Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Thrombospondin 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580122
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580122 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580122 Supplier Invitrogen™ Supplier No. PA580122
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human HepG2 whole cell, human K562 whole cell, rat C6 whole cell, mouse NIH/3T3 whole cell.

Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thrombospondin 1
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene THBS1
Gene Accession No. P07996
Gene Alias glycoprotein G; tbsp1; THBS; Thbs1; Thbs-1; thrombospondin; thrombospondin 1; thrombospondin-1; thrombospondin-1p180; thrombospondin-p50; ThrombospondinTHBS1; TSP; TSP1; TSP-1
Gene Symbols THBS1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the N-terminus of human THBS1 (58-86aa AFRIEDANLIPPVPDDKFQDLVDAVRAEK).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7057
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.