Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thrombospondin 1 Polyclonal Antibody

Thrombospondin 1 Polyclonal Antibody
Supplier: Thermo Scientific PA5142448
This item is not returnable.
View return policy
Description
Thrombospondin 1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
| Thrombospondin 1 | |
| Polyclonal | |
| Unconjugated | |
| THBS1 | |
| glycoprotein G; tbsp1; THBS; Thbs1; Thbs-1; thrombospondin; thrombospondin 1; thrombospondin-1; thrombospondin-1p180; thrombospondin-p50; ThrombospondinTHBS1; TSP; TSP1; TSP-1 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 21825, 7057 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P07996, P35441 | |
| THBS1 | |
| Peptide with sequence NRIPESGGDNSVFD-C (aa 19-32). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction