Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombospondin-2 Antibody (MM0568-9T19), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB211923488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB211923488 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB211923488 Supplier Novus Biologicals Supplier No. NBP211923AF488
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Thrombospondin-2 Monoclonal specifically detects Thrombospondin-2 in Human samples. It is validated for Western Blot.

Specifications

Antigen Thrombospondin-2
Applications Western Blot
Classification Monoclonal
Clone MM0568-9T19
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias thrombospondin 2, TSP2thrombospondin-2
Gene Symbols THBS2
Host Species Mouse
Immunogen Human recombinant Thrombospondin-2
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7058
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.