Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thrombospondin-2 Antibody (MM0568-9T19), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211923AF594
Description
Thrombospondin-2 Monoclonal specifically detects Thrombospondin-2 in Human samples. It is validated for Western Blot.Specifications
Thrombospondin-2 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
THBS2 | |
Human recombinant Thrombospondin-2 | |
0.1 mL | |
7058.0 | |
Store at 4°C in the dark. |
Western Blot | |
MM0568-9T19 | |
Western Blot | |
thrombospondin 2, TSP2thrombospondin-2 | |
Mouse | |
Protein G purified | |
Primary | |
Human | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction