Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Thrombospondin 2 Monoclonal Antibody (GT147)
GREENER_CHOICE

Catalog No. PIMA542342
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542342 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542342 Supplier Invitrogen™ Supplier No. MA542342
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

The Thrombospondin proteins (TSP 1-5) compose a family of glycoproteins that are involved in cell-to-cell and cell-to-matrix signaling. These extracellular, cell-surface proteins form complexes of both homo- and hetero-multimers. Thrombospondins play a role in development, aggregation of platelets, adhesion and migration of cells and progression of cells through the growth cycle. Thrombospondin 1 is released from platelets in response to Thrombin stimulation and is a transient component of the extracellular matrix of developing and repairing tissues. Thrombospondin 2 shares a high degree of homology with Thrombospondin 1, and is thought to have overlapping but unique functions. Thrombospondin 3 is a developmentally regulated heparin-binding protein. Thrombospondin 4 is neuronally expressed and stimulates neurite outgrowth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thrombospondin 2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone GT147
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and no preservative
Gene THBS2
Gene Accession No. P35442
Gene Alias Thbs2; Thbs-2; thrombospondin 2; Thrombospondin2; thrombospondin-2; TSP2; TSP-2; TSP2thrombospondin-2
Gene Symbols THBS2
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human Thrombospondin 2. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7058
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.