Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Thrombospondin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565381
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565381 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565381 Supplier Invitrogen™ Supplier No. PA565381
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HVTTEYVGPSS ERRPEVCERS CEELGNMVQE LSGLHVLVNQ LSENLKRVSN DNQFLWELIG GPPKTRNMSA CWQDGRFFAE NETWVVDSCT TCTCKKFKTI CHQITCP Highest antigen sequence identity to the following orthologs - mouse 76%, rat 75%.

The Thrombospondin proteins (TSP 1-5) compose a family of glycoproteins that are involved in cell-to-cell and cell-to-matrix signaling. These extracellular, cell-surface proteins form complexes of both homo- and hetero-multimers. Thrombospondins play a role in development, aggregation of platelets, adhesion and migration of cells and progression of cells through the growth cycle. Thrombospondin 1 is released from platelets in response to Thrombin stimulation and is a transient component of the extracellular matrix of developing and repairing tissues. Thrombospondin 2 shares a high degree of homology with Thrombospondin 1, and is thought to have overlapping but unique functions. Thrombospondin 3 is a developmentally regulated heparin-binding protein. Thrombospondin 4 is neuronally expressed and stimulates neurite outgrowth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thrombospondin 2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene THBS2
Gene Accession No. P35442
Gene Alias Thbs2; Thbs-2; thrombospondin 2; Thrombospondin2; thrombospondin-2; TSP2; TSP-2; TSP2thrombospondin-2
Gene Symbols THBS2
Host Species Rabbit
Immunogen Recombinant Human Thrombospondin 2. Recombinant protein control fragment (Product #RP-106064).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7058
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.