Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Thrombospondin 3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584922
Description
Immunogen sequence: VGALSECPFQ GDESIHSAVT NALHSILGEQ TKALVTQLTL FNQILVELRD DIRDQVKEMS LIRNTIMECQ VCGFHEQRS.
Adhesive glycoprotein that mediates cell-to-cell and cell-to-matrix interactions. Can bind to fibrinogen, fibronectin, laminin and type V collagen.
Specifications
Thrombospondin 3 | |
Polyclonal | |
Unconjugated | |
THBS3 | |
THBS3; Thbs-3; thrombospondin 3; thrombospondin-3; Tsp3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7059 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P49746 | |
THBS3 | |
Recombinant protein corresponding to Human THBS3. Recombinant protein control fragment (Product #RP-108611). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction