Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Thrombospondin 4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568467
Description
Predicted to react with Human samples.
Thrombospondin-4 (THBS4) belongs to the thrombospondin protein family, a family of adhesive calcium-binding glycoproteins that mediate cell-to-cell and cell-to-matrix interactions. Studies suggest that this protein may be involved in local signaling in the developing and adult nervous system. THBS4 is widely expressed and is abundant in atherosclerotic lesions and in areas prone to the development of lesions. It has been suggested to play an important role in the recruitment of macrophages and the local regulation of inflammation associated with atherogenesis. THBS4 has also been suggested as a putative tumor-suppressor protein in colorectal cancer.
Specifications
Thrombospondin 4 | |
Polyclonal | |
Unconjugated | |
THBS4 | |
THBS4; thrombospondin 4; thrombospondin-4; TSP4; TSP-4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7060 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P35443 | |
THBS4 | |
Synthesized peptide derived from human Thrombospondin 4 [aa 551-660]. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction