Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Thrombospondin 4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143371
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143371 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143371 Supplier Invitrogen™ Supplier No. PA5143371
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial. Positive control: zebrafish muscle.

Thrombospondin-4 (THBS4) belongs to the thrombospondin protein family, a family of adhesive calcium-binding glycoproteins that mediate cell-to-cell and cell-to-matrix interactions. Studies suggest that this protein may be involved in local signaling in the developing and adult nervous system. THBS4 is widely expressed and is abundant in atherosclerotic lesions and in areas prone to the development of lesions. It has been suggested to play an important role in the recruitment of macrophages and the local regulation of inflammation associated with atherogenesis. THBS4 has also been suggested as a putative tumor-suppressor protein in colorectal cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thrombospondin 4
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Alias THBS4; thrombospondin 4; thrombospondin-4; TSP4; TSP-4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of zebrafish Thbs4b. The exact sequence is proprietary.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.