Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Thromboxane synthase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596025
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596025 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596025 Supplier Invitrogen™ Supplier No. PA596025
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: A-549, HT-29, U-937, NCI-H460; Cellular Location: Endoplasmic reticulum membrane, Multi-pass membrane protein Immunogen sequence: TYLLATNPDC QEKLLREVDV FKEKHMAPEF CSLEEGLPYL DMVIAETLRM YPPAFRFTRE AAQDCEVLGQ RIPAGAVLEM AVGALHHDPE HWPSPETFNP ERFTAEARQQ HRPFTYLPFG AGPRSCLGVR LGLLEVKLTL LHVLHKFRFQ ACPETQVPLQ LESKSALGPK NGVYIKIVSR.

Thromboxane synthase encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. However, this protein is considered a member of the cytochrome P450 superfamily on the basis of sequence similarity rather than functional similarity. This endoplasmic reticulum membrane protein catalyzes the conversion of prostglandin H2 to thromboxane A2, a potent vasoconstrictor and inducer of platelet aggregation. The enzyme plays a role in several pathophysiological processes including hemostasis, cardiovascular disease, and stroke. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thromboxane synthase
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene TBXAS1
Gene Accession No. P24557, P36423, P49430
Gene Alias BDPLT14; CYP5; Cyp5a1; cytochrome P450 5A1; cytochrome P450, family 5, subfamily A, polypeptide 1; FLJ52771; GHOSAL; platelet, cytochrome P450, subfamily V; similar to Thromboxane-A synthase (TXA synthase) (TXS); Tbxas1; THAS; ThromboxA ane synthase 1; thromboxane A synthase 1; thromboxane A synthase 1 (platelet); thromboxane A synthase 1 (platelet, cytochrome P450, family 5, subfamily A); thromboxane A synthase 1, platelet; thromboxane synthase; thromboxane-A synthase; TS; TXA synthase; TXAS; TXS
Gene Symbols TBXAS1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 355-534 of human Thromboxane synthase (NP_0010522).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21391, 24886, 6916
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.