Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THRSP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA548103
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA548103 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA548103 Supplier Invitrogen™ Supplier No. PA548103
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.

The protein encoded by this gene is similar to the gene product of S14, a rat gene whose expression is limited to liver and adipose tissue and is controlled by nutritional and hormonal factors. This gene has been shown to be expressed in liver and adipocytes, particularly in lipomatous modules. It is also found to be expressed in lipogenic breast cancers, which suggests a role in controlling tumor lipid metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THRSP
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene THRSP
Gene Accession No. Q62264
Gene Alias lipogenic protein 1; Lipogenic protein S14; Lpgp; LPGP1; S14; S14MGC21659; Spot 14 protein; SPOT14; SPOT14 homolog; THRP; Thrsp; thyroid hormone responsive; thyroid hormone responsive (SPOT14 homolog, rat); thyroid hormone responsive SPOT14; thyroid hormone responsive SPOT14 homolog; thyroid hormone-inducible hepatic protein
Gene Symbols THRSP
Host Species Goat
Immunogen E. coli-derived recombinant mouse THRSP Gly47-Leu150.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21835
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.