Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THRSP Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987460
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7460 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7460 Supplier Bioss Supplier No. BS6738R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is similar to the gene product of S14, a rat gene whose expression is limited to liver and adipose tissue and is controlled by nutritional and hormonal factors. This gene has been shown to be expressed in liver and adipocytes, particularly in lipomatous modules. It is also found to be expressed in lipogenic breast cancers, which suggests a role in controlling tumor lipid metabolism.

Specifications

Antigen THRSP
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene THRSP
Gene Accession No. Q92748
Gene Alias lipogenic protein 1; Lipogenic protein S14; Lpgp; LPGP1; S14; S14MGC21659; Spot 14 protein; SPOT14; SPOT14 homolog; THRP; Thrsp; thyroid hormone responsive; thyroid hormone responsive (SPOT14 homolog, rat); thyroid hormone responsive SPOT14; thyroid hormone responsive SPOT14 homolog; Thyroid hormone-inducible hepatic protein
Gene Symbols THRSP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human THRSP.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7069
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.