Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
THSD1 Antibody (541213), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB5178MFV500
Description
THSD1 Monoclonal antibody specifically detects THSD1 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| THSD1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer | |
| 55901 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 541213 | |
| Flow Cytometry, CyTOF-ready | |
| thrombospondin type-1 domain-containing protein 1, thrombospondin, type I, domain 1, thrombospondin, type I, domain containing 1,4833423O18Rik, TMTSPMGC74971, Transmembrane molecule with thrombospondin module, UNQ3010 | |
| Mouse myeloma cell line NS0-derived recombinant human THSD1 isoform 2, Glu25-Ile361, Accession # NP_954872 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction