Learn More
Invitrogen™ THSD1 Monoclonal Antibody (541213)

Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | THSD1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 541213 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | THSD1 |
| Gene Accession No. | Q9NS62 |
| Gene Alias | 4833423O18Rik; LOW QUALITY PROTEIN: thrombospondin type-1 domain-containing protein 1; thrombospondin type 1 domain containing 1; thrombospondin type I domain-containing 1 isoform 1; thrombospondin type-1 domain-containing protein 1; thrombospondin, type I, domain 1; thrombospondin, type I, domain containing 1; Thsd1; TMTSP; Transmembrane molecule with thrombospondin module; UNQ3010; UNQ3010/PRO9769 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.