Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THSD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582464
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582464 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582464 Supplier Invitrogen™ Supplier No. PA582464
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VPPEDDASGS ESFQSNAQKI IPPLFSYRLA QQQLKEMKKK GLTETTKVYH VSQSPLTDTA IDAAPSAPLD LESPEEAAAN KFRIKSPFPE QPAVSAGERP PSRLDLNVTQ Highest antigen sequence indentity to the following orthologs: Mouse - 82%, Rat - 82%.

The protein encoded by this gene contains a type 1 thrombospondin domain, which is found in a number of proteins involved in the complement pathway, as well as in extracellular matrix proteins. Alternatively spliced transcript variants encoding different isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THSD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene THSD1
Gene Accession No. Q9NS62
Gene Alias 4833423O18Rik; LOW QUALITY PROTEIN: thrombospondin type-1 domain-containing protein 1; thrombospondin type 1 domain containing 1; thrombospondin type I domain-containing 1 isoform 1; thrombospondin type-1 domain-containing protein 1; thrombospondin, type I, domain 1; thrombospondin, type I, domain containing 1; Thsd1; TMTSP; Transmembrane molecule with thrombospondin module; UNQ3010; UNQ3010/PRO9769
Gene Symbols THSD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human THSD1. Recombinant protein control fragment (Product #RP-90243).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55901
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.