Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THSD1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987789
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7789 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7789 Supplier Bioss Supplier No. BS7508R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene contains a type 1 thrombospondin domain, which is found in a number of proteins involved in the complement pathway, as well as in extracellular matrix proteins. Alternatively spliced transcript variants encoding different isoforms have been observed for this gene.

Specifications

Antigen THSD1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene THSD1
Gene Accession No. Q9NS62
Gene Alias 4833423O18Rik; BOS_12369; thrombospondin type 1 domain containing 1; thrombospondin type I domain-containing 1; Thrombospondin type-1 domain-containing protein 1; thrombospondin, type I, domain 1; thrombospondin, type I, domain containing 1; THSD1; Tmtsp; Transmembrane molecule with thrombospondin module; UNQ3010; UNQ3010/PRO9769
Gene Symbols THSD1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human THSD1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55901
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.