Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THSD7B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566037
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566037 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566037 Supplier Invitrogen™ Supplier No. PA566037
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PTQGEGRPCPT ELTQEKTCPV TPCYSWVLGN WSACKLEGGD CGEGVQIRSL SCMVHSGSIS HAAGRVEDAL CGEMPFQDSI LKQLCSV Highest antigen sequence identity to the following orthologs - mouse 84%, rat 82%.

THSD7B is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THSD7B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene THSD7B
Gene Accession No. Q9C0I4
Gene Alias KIAA1679; thrombospondin type 1 domain containing 7B; thrombospondin type-1 domain-containing protein 7B; thrombospondin, type I, domain containing 7B; THSD7B
Gene Symbols THSD7B
Host Species Rabbit
Immunogen Recombinant Human THSD7B. Recombinant protein control fragment (Product #RP-106727).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80731
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.