Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ THSD7B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562404
Description
Immunogen sequence: VLMESTGPAG HCPHLVESVP CEDPMCYRWL ASEGICFPDH GKCGLGHRIL KAVCQNDRGE DVSGSLCPVP PPPERKSCEI PCRMDCVLSE W Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 90%.
THSD7B is a protein coding gene.
Specifications
THSD7B | |
Polyclonal | |
Unconjugated | |
THSD7B | |
KIAA1679; thrombospondin type 1 domain containing 7B; thrombospondin type-1 domain-containing protein 7B; thrombospondin, type I, domain containing 7B; THSD7B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
80731 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9C0I4 | |
THSD7B | |
Recombinant protein corresponding to Human THSD7B. Recombinant protein control fragment (Product #RP-101354). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction