Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ THTPA Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555998
Description
Immunogen sequence: GAAGVLGPHT EYKELTAEPT IVAQLCKVLR ADGLGAGDVA AVLGPLGLQE VASFVTKRSA WKLVLLGADE EEPQLRVDLD TADFGYAVGE VEALVHEEAE VPTALEKIHR LSSMLGVP Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 65%.
THTPA encodes an enzyme which catalyzes the biosynthesis of thiamine disphophate (vitamin B1) by hydrolysis of thiamine triphosphate. Alternative splicing results in multiple transcript variants.
Specifications
| THTPA | |
| Polyclonal | |
| Unconjugated | |
| THTPA | |
| AW122966; thiamine triphosphatase; thiamine-triphosphatase; THTP; Thtpa; ThTPase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 79178 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BU02 | |
| THTPA | |
| Recombinant protein corresponding to Human THTPA. Recombinant protein control fragment (Product #RP-89100). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction