Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thulium(III) chloride hydrate, REacton™, 99.99% (REO)

Catalog No. p-4904552
Click to view available options
Quantity:
0.5 g
2 g
10 g

CAS: 19423-86-0 | Cl3Tm | 275.28 g/mol

Thulium(III) chloride hydrate is used extremely as a rare metal to study the metal science of thulium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19423-86-0
Molecular Formula Cl3Tm
Molecular Weight (g/mol) 275.28
MDL Number MFCD00011283
InChI Key ILOTUXNTERMOJL-UHFFFAOYSA-K
IUPAC Name thulium(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[Tm+3]

Specifications

Color Green
Physical Form Wet Crystalline Aggregates
Quantity 0.5 g
Assay Percent Range 99.99% (REO)
Solubility Information Soluble in water.
Formula Weight 275.29 (Anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Thulium(III) chloride hydrate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-904-9
RTECSNumber XP0525000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.