Thulium(III) chloride hydrate, 99.99% (REO), REacton™, Alfa Aesar™

Manufacturer: Alfa Aesar 1120104

Quantity 2g

 View more versions of this product

Catalog No. AA1120104

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 19423-86-0
    Physical Form Wet Crystalline Aggregates
    Assay Percent Range 99.99% (REO)
    Molecular Formula TmCl3·xH2O
    Formula Weight 275.29
    MDL Number MFCD00011283

    Specifications

    Formula Weight 275.29
    Quantity 2g
    Color Green
    Physical Form Wet Crystalline Aggregates
    Odor Odorless
    Assay Percent Range 99.99% (REO)
    Molecular Formula TmCl3·xH2O
    MDL Number MFCD00011283
    CAS 19423-86-0

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit