Learn More
Thulium(III) oxide, REacton™, 99.9% (REO)
Description
Thulium oxide is an excellent thulium source which finds use in glass, optical and ceramic applications. Further, it is used in the manufacturing of portable X-ray transmission device. In addition to this, it finds use as a nuclear reactor control material. Nanostructured thulium oxide acts as an efficient biosensor in the field of medicinal chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 2425°C |
Physical Form | Powder |
Quantity | 10 g |
Assay Percent Range | 99.9% (REO) |
Solubility Information | Dissolves slowly in strong acids.Slightly soluble in strong acids. Insoluble in water. |
Formula Weight | 385.87 |
Chemical Name or Material | Thulium(III) oxide |
Safety and Handling
EINECSNumber | 234-851-6 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.