Thulium(III) oxide, REacton™, 99.9% (REO), Alfa Aesar™

Manufacturer: Alfa Aesar 11198-18

Quantity 50g

 View more versions of this product

Catalog No. AA1119818

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12036-44-1
    Molecular Formula O3Tm2
    Linear Formula Tm2O3
    Formula Weight 385.87g/mol
    MDL Number MFCD00011285
    Merck Index 14,9394
    Chemical Name or Material Thulium(III) oxide, REacton™, (REO)

    Specifications

    Color Pale Green
    Melting Point 2425°C
    Quantity 50g
    CAS 12036-44-1
    Physical Form Powder
    Molecular Formula O3Tm2
    Linear Formula Tm2O3
    Formula Weight 385.87g/mol
    MDL Number MFCD00011285
    Merck Index 14,9394
    Solubility Information Dissolves slowly in strong acids
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Chemical Name or Material Thulium(III) oxide, REacton™, (REO)
    Assay 99.9%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit