Learn More
Thulium(III) oxide, REacton™, 99.997% (REO)
Description
Thulium(III) oxide when irradiated in a nuclear reactor, thulium produces an isotope that emits x-rays. A button of this isotope is used to make a lightweight, portable x-ray machine for medical use. It is also used in lasers with surgical applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 2425°C |
| Color | Green |
| Physical Form | Powder |
| Quantity | 1 g |
| Linear Formula | Tm2O3 |
| Merck Index | 14,9394 |
| Solubility Information | Insoluble in water. Slightly soluble in acids. |
| Formula Weight | 385.87 |
| Percent Purity | 99.997% |
| Grade | Reacton™ |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 234-851-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.