Description
- Features a dual tab
- Available in various colors and sizes
- 1000 per case
Specifications
Specifications
| Closure Size | 12/13 mm |
| Liner Material | Unlined |
| Autoclavable | Not Autoclavable |
| Material | Plastic |
| Color | White |
| No. per Case | 1000 |
| Sterility | Non-sterile |
| Disposable | Single-use |
| Fit for Purpose | PCR Ready |