Learn More
Fisherbrand™ Thumb Cap
Catalog No. 23001706 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Gray
Green
Lavender
White
Yellow
Closure Size:
12/13 mm
13 mm
16 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Size
23-001-706 Yellow 12/13 mm
23-001-702 Green 16 mm
23-001-703 Gray 12/13 mm
23-001-705 White 16 mm
23-001-708 Lavender 13 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 23-001-706
Add to Cart
Add to Cart
Colored thumb cap with dual tab.
  • Features a dual tab
  • Available in various colors and sizes
  • 1000 per case
Specifications
Closure Size 12/13 mm
Liner Material Unlined
Autoclavable Not Autoclavable
Material Plastic
Color Yellow
No. per Case 1000
Sterility Non-sterile
Disposable Single-use
Fit for Purpose PCR Ready