Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
THUMPD1 Antibody (OTI1H6), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274512F
Description
THUMPD1 Monoclonal specifically detects THUMPD1 in Human, Mouse, Monkey samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
THUMPD1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
THUMPD1 | |
Full length recombinant protein of human THUMPD1 (NP_060206) produced in HEK293T cells. | |
0.1 mL | |
55623.0 | |
Store at 4°C in the dark. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI1H6 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
DKFZp686C1054, FLJ20274, THUMP domain containing 1, THUMP domain-containing protein 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction