Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THUMPD1 Monoclonal Antibody (4A11)
GREENER_CHOICE

Catalog No. PIMA524483
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524483 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524483 Supplier Invitrogen™ Supplier No. MA524483
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MAAPAQQTTQ PGGGKRKGKA QYVLAKRARR CDAGGPRQLE PGLQGILITC NMNERKCVEE AYSLLNEYGD DMYGPEKFTD KDQQPSGSEG EDDDAEAALK KEVGDIKAST EMRLRRFQSV ESGANNVVFI RTLGIEPEKL VHHILQDMYK TKKKKTRVIL RMLPISGTCK AFLEDMKKYA ETFLEPWFKA PNKGTFQIVY KSRNNSHVNR EEVIRELAGI VCTLNSENKV DLTNPQYTVV VEIIKAVCCL SVVKDYMLFR KYNLQEVVKS PKDPSQLNSK QGNGKEAKLE SADKSDQNNT AEGKNNQQVP ENTEELGQTK PTSNPQVVNE GGAKPELASQ ATEGSKSNEN DFS.

Functions as a tRNA-binding adapter to mediate NAT10-dependent tRNA acetylation (PubMed:25653167). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen THUMPD1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 4A11
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Thumpd1
Gene Accession No. Q9NXG2
Gene Alias 6330575P11Rik; EGK_12570; Tan1; THUMP domain containing 1; THUMP domain-containing protein 1; THUMPD1
Gene Symbols Thumpd1
Host Species Mouse
Immunogen THUMPD1 (AAH00448, 1 a.a. ~ 353 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55623
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.