Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THUMPD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555714
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555714 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555714 Supplier Invitrogen™ Supplier No. PA555714
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RNNSHVNREE VIRELAGIVC TLNSENKVDL TNPQYTVVVE IIKAVCCLSV VKDYMLFRKY NLQEVVKSPK DPSQLNSKQG NGKEAKLE Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 84%.

Functions as a tRNA-binding adapter to mediate NAT10-dependent tRNA acetylation (PubMed:25653167). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen THUMPD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Thumpd1
Gene Accession No. Q9NXG2
Gene Alias 6330575P11Rik; EGK_12570; Tan1; THUMP domain containing 1; THUMP domain-containing protein 1; THUMPD1
Gene Symbols Thumpd1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human THUMPD1. Recombinant protein control fragment (Product #RP-90699).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55623
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.