Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THUMPD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588232
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588232 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588232 Supplier Invitrogen™ Supplier No. PA588232
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IVLLLSEDHH RRLTDCKESN IPFNSKDSHT DEPGIKKCLN PEEKTGAFKT ASTSFEASNH KFLDRMSPFG SLVPVECYKV SLGKTDAFIC KYKKSHSSGL; Positive Samples: A-549, HeLa, HT-29, 293T, Mouse brain, Rat liver.

THUMPD2 (THUMP Domain Containing 2) is a Protein Coding gene. Diseases associated with THUMPD2 include Retinitis Pigmentosa 11. Gene Ontology (GO) annotations related to this gene include RNA binding and methyltransferase activity. An important paralog of this gene is THUMPD3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen THUMPD2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Thumpd2
Gene Accession No. Q9BTF0, Q9CZB3
Gene Alias 2810025A12Rik; C2orf8; SAM-dependent methyltransferase; thum2; THUMP domain containing 2; THUMP domain-containing protein 2; THUMPD2
Gene Symbols Thumpd2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 404-503 of human THUMPD2 (NP_0795402).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 313851, 72167, 80745
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.