Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ THUMPD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557554
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557554 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557554 Supplier Invitrogen™ Supplier No. PA557554
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NLPLRTGSVD IIVTDLPFGK RMGSKKRNWN LYPACLREMS RVCTPTTGRA VLLTQDTKCF TKALSGMRHV WRKVDTVWVN VGGLRAAVYV LIRTPQAFVH P Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 88%.

THUMPD3 a protein of unknown function. Its domain structure suggests that it is a methylase. It contains a THUMP domain (an RNA-binding domain) and a UPF0020 domain (putative methylase).
TRUSTED_SUSTAINABILITY

Specifications

Antigen THUMPD3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene THUMPD3
Gene Accession No. Q9BV44
Gene Alias AW556087; gene trap ROSA 26 antisense, Philippe Soriano; Gt(ROSA)26asSor; Gtrosa26as; GtROSA26asSor; RGD1560852; thum3; THUMP domain containing 3; THUMP domain-containing protein 3; THUMPD3
Gene Symbols THUMPD3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human THUMPD3. Recombinant protein control fragment (Product #RP-96338).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25917
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.