Learn More
Invitrogen™ THUMPD3 Polyclonal Antibody

Description
Immunogen sequence: MCDIEEATNQ LLDVNLHENQ KSVQVTESDL GSESELLVTI GATVPTGFEQ TAADEVREKL GSSCKISRDR GKIYFVISVE SLAQVHCLRS VDNLFVVVQE FQDYQFKQTK EEVLKDFEDL AGKLPWSNPL KVWKINASFK KKKAKRKKIN QNSSKEKINN GQEVKIDQRN VKKEFTSHAL DSHILDYYEN PAIKEDVSTL IGDDLASCKD ETDESSKEET EPQVLKFRVT CNRAGEKHCF TSNEAARDFG; Positive Samples: LO2, A-549, MCF-7.
Specifications
Specifications
| Antigen | THUMPD3 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 3.95 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | THUMPD3 |
| Gene Accession No. | P97770, Q9BV44 |
| Gene Alias | AW556087; gene trap ROSA 26 antisense, Philippe Soriano; Gt(ROSA)26asSor; Gtrosa26as; GtROSA26asSor; RGD1560852; thum3; THUMP domain containing 3; THUMP domain-containing protein 3; THUMPD3 |
| Gene Symbols | THUMPD3 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.