Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thy 1.1 (CD90) Mouse anti-Rat, Mouse, Clone: T11D7e, MilliporeSigma™

Catalog No. MABF1961MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1961MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF1961MI Supplier MilliporeSigma Supplier No. MABF1961
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This mouse monoclonal Anti-Thy 1.1 (CD90), clone T11D7e CD90.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thy 1.1 (CD90)
Applications Flow Cytometry
Classification Monoclonal
Clone T11D7e
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Thy1,Thy-1
Host Species Mouse
Immunogen Xenogenic rat thymocytes.
Purification Method Purified by ion-exchange chromatography
Quantity 200 μL
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_036805
Test Specificity Clone T11D7e detects Thy-1.1 (CD90) in rat thymocytes.
Target Species Rat, Mouse
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.