Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thy1 Rat anti-Mouse, Clone: 5A4D10, Abnova™

Catalog No. 89035666 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
40 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-666 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-666 Supplier Abnova Corporation Supplier No. MAB14230

Rat Monoclonal Antibody

Rat monoclonal antibody raised against full length recombinant mouse Thy1.

  • High specificity
  • High reactivity

Specifications

Antigen Thy1
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone 5A4D10
Conjugate Unconjugated
Description Rat monoclonal antibody raised against full length recombinant mouse Thy1.
Dilution ELISA (0.05-0.2 ug/mL) Flow Cytometry (1-3 ug x 106 cells) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS, pH 7.4 (0.02% sodium azide).
Gene Accession No. P01831
Gene Alias CD90/T25/Thy-1/Thy-1.2/Thy1.1/Thy1.2
Gene Symbols Thy1
Host Species Rat
Immunogen Recombinant protein corresponding to full length mouse Thy1.
Purification Method Protein G purification
Quantity 40 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21838
Target Species Mouse
Content And Storage Store at -20°C on dry atmosphere, lyophilized antibodies are stable.
After reconstitution with deionized water, store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.