Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thymidine glycol Rabbit anti-Human, Clone: B29, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP311041100UG
Description
Thymidine glycol Monoclonal specifically detects Thymidine glycol in Human samples. It is validated for ELISA, Immunohistochemistry.Specifications
Thymidine glycol | |
Monoclonal | |
1 mg/ml | |
ELISA, Immunohistochemistry 1:100 | |
Rabbit | |
Affinity purified | |
RUO | |
Human | |
Purified |
ELISA, Immunohistochemistry | |
B29 | |
Unconjugated | |
PBS (pH 7.4), 50% Glycerol | |
Thymidine glycol polymer | |
100 μg | |
Primary | |
Store at -20°C. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction