Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thymidine Kinase 1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10060644AF488
Description
Thymidine Kinase 1 Polyclonal antibody specifically detects Thymidine Kinase 1 in Human samples. It is validated for Western BlotSpecifications
Thymidine Kinase 1 | |
Polyclonal | |
Western Blot | |
EC 2.7.1.21, thymidine kinase 1 soluble isoform, thymidine kinase 1, soluble, thymidine kinase, cytosolic, thymidine kinase-1, TK2 | |
The immunogen recognized by this antibody maps to a region between residue 205 and 234 of human thymidine kinase 1 using the numbering given in entry NP_003249.1 (GeneID 7083). | |
0.1 mL | |
DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers | |
7083 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction