Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymidine Kinase 1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP220626

Catalog No. NBP220626


Only null left
Add to Cart

Description

Description

Thymidine Kinase 1 Polyclonal specifically detects Thymidine Kinase 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Thymidine Kinase 1
Polyclonal
Unconjugated
PBS, 20% Glycerol (pH7). with 0.025% Proclin 300
EC 2.7.1.21, thymidine kinase 1 soluble isoform, thymidine kinase 1, soluble, thymidine kinase, cytosolic, thymidine kinase-1, TK2
Rabbit
25 kDa
0.1 mL
DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers
7083
Human, Mouse, Rat
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
1mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
P04183
TK1
Recombinant protein encompassing a sequence within the center region of human Thymidine Kinase 1. The exact sequence is proprietary.
Affinity Purified
RUO
Primary
Species reactivity based on sequence identity: Chicken (83%), Xenopus tropicalis (81%).
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only