Learn More
Description
Host Species: Mouse
Clone: P-GF.44C
Isotype: IgG1
Species Reactivity: Human, Mouse and Rat. Others not known.
Epitope: Not determined
Immunogen: Recombinant full-length human thymidine phosphorylase (TP/PD-ECGF) protein
Molecular Weight: 55kDa (reduced)
Positive Control: HUVEC cells, Breast CA
Cellular Localization: Cytoplasmic and nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting (Not verified)
- Neutralizes Enzymatic Activity of TP/PD-ECGF (Not verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Thymidine Phosphorylase/PD-ECGF Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | P-GF.44C |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant full length human thymidine phosphorylase (TP/PD-ECGF) protein |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.