Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymidine Phosphorylase/PD-ECGF Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS499PABX Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS499PABX 200 μL Concentrated 1mg/mL
MS499P0 100 μL Concentrated 200μg/mL
MS499P1 500 μL Concentrated 200μg/mL
MS499P 1 mL Concentrated 200μg/mL
MS499P1ABX 100 μL Concentrated 1mg/mL
MS499R1 1 mL Prediluted
MS499R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS499PABX Supplier Epredia Supplier No. MS499PABX

Provide accurate, reproducible results with the Epredia™ Thymidine Phosphorylase/PD-ECGF Ab-1, Mouse Monoclonal Antibody.

Platelet-derived endothelial growth factor (PD-ECGF), same as thymidine phosphorylase (TP) or gliostatin. In the presence of inorganic orthophosphate, it catalyses the reversible phospholytic cleavage of thymidine and deoxyuridine to their corresponding bases and 2-deoxyribose-1-phosphate. It is both chemotactic and mitogenic for endothelial cells and a non-heparin binding angiogenic factor present in platelets. It is also involved in transformation of fluoropyrimidines, cytotoxic agents used in the treatment of a variety of malignancies, into active cytotoxic metabolites.

Host Species: Mouse

Clone: P-GF.44C

Isotype: IgG1

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: Not determined

Immunogen: Recombinant full-length human thymidine phosphorylase (TP/PD-ECGF) protein

Molecular Weight: 55kDa (reduced)

Positive Control: HUVEC cells, Breast CA

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Neutralizes Enzymatic Activity of TP/PD-ECGF (Not verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Thymidine Phosphorylase/PD-ECGF Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone P-GF.44C
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant full length human thymidine phosphorylase (TP/PD-ECGF) protein
Quantity 200 μL
Regulatory Status RUO
Research Discipline Angiogenesis
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.