Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thymidine Phosphorylase Polyclonal Antibody
Thymidine Phosphorylase Polyclonal Antibody
Supplier: Thermo Scientific PA588060
Description
Thymidine Phosphorylase Polyclonal Antibody for Western Blot, ICC/IF

Specifications
Thymidine Phosphorylase | |
Polyclonal | |
Unconjugated | |
TYMP | |
2900072D10Rik; ECGF; Ecgf1; endothelial cell growth factor 1 (platelet-derived); gliostatin; hPD-ECGF; MEDPS1; MNGIE; MTDPS1; OTTHUMP00000196767; OTTHUMP00000196770; PD TP; PDECGF; PD-ECGF; Pdgfec; platelet derived growth factor, endothelial cell; Platelet-derived endothelial cell growth factor; TdRPase; thymidine phosphorylase; TP; TYMP | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1890 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.98 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P19971 | |
TYMP | |
Recombinant fusion protein containing a sequence corresponding to amino acids 11-482 of TYMP (NP_0019441). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction