Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thymidylate Synthase Antibody (TMS715), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234725MFV500
Description
Thymidylate Synthase Monoclonal antibody specifically detects Thymidylate Synthase in Human samples. It is validated for Flow Cytometry, Flow (Intracellular)Specifications
Thymidylate Synthase | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
TMS715 | |
Flow Cytometry, Flow (Intracellular) | |
EC 2.1.1.45, HsT422, MGC88736, thymidylate synthase, thymidylate synthetase, TMSTsase, TSase, TSHST422 | |
Recombinant full-length human Thymidylate Synthase protein. (Uniprot: P04818 ) | |
0.1 mL | |
Cancer, Cell Cycle and Replication, DNA Repair, DNA replication Transcription Translation and Splicing, Transcription Factors and Regulators, Translation Control | |
7298 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction