Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Thymidylate Synthase Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA522263

Catalog No. PIPA522263


Only null left
Add to Cart

Description

Description

Recommended positive controls: A549, H1299, HCT116. Predicted reactivity: Mouse (94%), Rat (95%), Zebrafish (88%), Xenopus laevis (85%), Dog (96%), Pig (96%), Rhesus Monkey (98%), Chimpanzee (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Thymidylate Synthase (TS) converts deoxyuridine monophosphate (dUMP) to deoxythymidine monophosphate (dTMP), which is essential for DNA biosynthesis. TS is a critical target for the fluoropyrimidines, an important group of antineoplastic drugs that are widely used in the treatment of solid tumors. Both 5-FU and fluorodeoxyuridine are converted in tumor cells to FdUMP which inactivates TS by formation of a ternary covalent complex in the presence of the folate cofactor 5,10-methylenetetrahydrofolate. Expression of TS protein is associated with response to 5-fluorouracil (5-FU) in human colorectal, gastric, head and neck, and breast carcinomas.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Thymidylate Synthase
Polyclonal
Unconjugated
TYMS
HST422; MGC88736; OK/SW-cl.29; Thymidylate synthase; thymidylate synthetase; TMS; TS; Tsase; Tyms; tysy
Rabbit
Antigen affinity chromatography
RUO
7298
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
P04818
TYMS
Recombinant protein encompassing a sequence within the center region of human Thymidylate synthase. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.