Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymidylate SynthaseSheep anti-Human, Mouse, Rat, Polyclonal, Rockland™
SDP

Catalog No. 501990211
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0211 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-0211 Supplier Rockland Immunochemicals Supplier No. 100601199
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antiserum is directed against human Thymidylate Synthase and is useful in determining its presence in various assays. Because inhibition of Thymidylate Synthase prevents DNA synthesis and cell proliferation, the enzyme is an important target for cancer chemotherapeutic drugs, specifically the fluoropyrimidine group of antineoplastic drugs used to treat solid tumors. In general, this antibody can detect antigen in a variety of human cells and tissues, as well as bacteria, African green monkey, rat and mouse. Somewhat lower dilutions may be required in some non-human cell lines. Anti-Thymidylate Synthase can detect Thymidylate Synthase by immunochemistry in proliferating cell cultures and tissues but does not stain nonproliferating cells. Normal colon mucosa shows weak staining; however, some colorectal cancer specimens show very strong staining.

Thymidylate Synthase (TS) converts deoxyuridine monophosphate (dUMP) to deoxythymidine monophosphate (dTMP), which is essential for DNA biosynthesis. TS is a critical target for the fluoropyrimidines, an important group of antineoplastic drugs that are widely used in the treatment of solid tumors. Both 5-FU and fluorodeoxyuridine are converted in tumor cells to FdUMP which inactivates TS by formation of a ternary covalent complex in the presence of the folate cofactor 5,10-methylenetetrahydrofolate. Expression of TS protein is associated with response to 5-fluorouracil (5-FU) in human colorectal, gastric, head and neck, and breast carcinomas.

Specifications

Antigen Thymidylate Synthase
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 85 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene TYMS
Gene Accession No. P04818, P07607, P45352
Gene Alias HST422; MGC88736; OK/SW-cl.29; Thymidylate synthase; thymidylate synthetase; TMS; TS; TSase; Tyms
Gene Symbols TYMS
Host Species Sheep
Immunogen This whole sheep serum was prepared by repeated immunizations with recombinant human Thymidylate Synthase (36 kDa) produced in E.coli.
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22171, 29261, 7298
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.