Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymol Blue, 0.08% (w/v) in Methanol, Ricca Chemical
SDP

Catalog No. 8420816 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
8420816 500 mL Natural Poly Bottle
842084 120 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 8420816 Supplier Ricca Chemical Company Supplier No. 8420.816
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 76-61-9 , 67-56-1
Molecular Formula C27H30O5S
Molecular Weight (g/mol) Mixture
InChI Key PRZSXZWFJHEZBJ-UHFFFAOYSA-N
PubChem CID 65565
IUPAC Name 4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenol
SMILES CC1=CC(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C=C4C)O)C(C)C)C(C)C)O

Specifications

CAS Min % 98.0, 0.1
CAS Max % 100.0, 0.1
Packaging Natural Poly Bottle
Color Dark Green
Quantity 500 mL
Physical Form Liquid
Percent Purity 0.08% W/V
Grade Indicator
Concentration or Composition (by Analyte or Components) 0.08% (w/v) in Methanol
Chemical Name or Material Thymol Blue
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.