Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Thymol Blue sodium salt

Catalog No. AAB2300718
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 62625-21-2 | C27H29NaO5S | 488.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62625-21-2
Molecular Formula C27H29NaO5S
Molecular Weight (g/mol) 488.57
MDL Number MFCD00151093
InChI Key OCMIKNBSIRSUPI-FRWNMSGJSA-M
Synonym Thymol Blue, water soluble
PubChem CID 23692293
IUPAC Name sodium;4-[3-(4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-5-methyl-2-propan-2-ylphenolate
SMILES [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O

Specifications

Melting Point ∼284°C (decomposition)
Beilstein 3861515
Formula Weight 488.58
Quantity 50 g
Chemical Name or Material Thymol Blue sodium salt
Hazard Category H500
EINECSNumber 263-650-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.